Learn More
Fritsch Milling And Sizing Inc Rotor 12 Ribs, Pure Titanium PULVERISETTE 14 Premium Line
SDP
Catalog No. 501533966
Encompass
Impact rotor for PULVERISETTE 14 premium line with 12 ribs and cooling fins, pure titanium, grinding of materials - feed size < 10 mm.
Catalog No. 50-153-3966 Supplier Fritsch Milling And Sizing Inc Supplier No. 14.4434.32
Add to Cart
Add to Cart
Request a Quote
Impact rotor for PULVERISETTE 14 premium line with 12 ribs and cooling fins, pure titanium, grinding of materials - feed size < 10 mm.