Learn More
Fritsch Milling And Sizing Inc Rotor 12 Ribs, Stainless Steel PULVERISETTE 14 Premium Line
SDP
Catalog No. 501533950
Encompass
The 12-ribs impact rotor for PULVERISETTE 14 premium line with cooling fins is suitable for the grinding of materials with feed size < 10 mm.
Catalog No. 50-153-3950 Supplier Fritsch Milling And Sizing Inc Supplier No. 14.4334.10
The 12-ribs impact rotor for PULVERISETTE 14 premium line with cooling fins is suitable for the grinding of materials with feed size < 10 mm.