Learn More
Fritsch Milling And Sizing Inc Rotor 24 Ribs, Pure Titanium PULVERISETTE 14 Premium Line
SDP
Catalog No. 501533967
Encompass
Impact rotor for PULVERISETTE 14 premium line with 24 ribs and cooling fins, pure titanium, for grinding fine materials - feed size < 5 mm.
Catalog No. 50-153-3967 Supplier Fritsch Milling And Sizing Inc Supplier No. 14.4437.32
Add to Cart
Add to Cart
Request a Quote
Impact rotor for PULVERISETTE 14 premium line with 24 ribs and cooling fins, pure titanium, for grinding fine materials - feed size < 5 mm.