Learn More
Fritsch Milling And Sizing Inc Rotor 24 Ribs, Stainless Steel PULVERISETTE 14 Premium Line
SDP
Supplier:  Fritsch Milling And Sizing Inc 14.4337.10
Encompass
The 24-ribs impact rotor for PULVERISETTE 14 premium line with cooling fins is suitable for the grinding of fine materials with a feed size of < 5 mm.
Catalog No. 50-153-3951