Learn More
Alkali Scientific Rotor, 44 X 1.5/2.0mL (15,000 rpm/ 21,379 xg)
SDP
Catalog No. 502389348
Encompass_Preferred
Rotor, 44 x 1.5/2.0mL (15,000 rpm/ 21,379 xg) compatible with Z216-M and Z216-MK Microcentrifuges.
Catalog No. 50-238-9348 Supplier Alkali Scientific Supplier No. RS6311
Rotor, 44 x 1.5/2.0mL (15,000 rpm/ 21,379 xg) compatible with Z216-M and Z216-MK Microcentrifuges.