Learn More
Fritsch Milling And Sizing Inc Rotor 6 Ribs, Pure Titanium PULVERISETTE 14 Premium Line
SDP
Catalog No. 501533965
Encompass
Impact rotor for PULVERISETTE 14 premium line with 6 ribs and cooling fins, pure titanium, fast comminution of fibrous substances - feed size < 15 mm.
Catalog No. 50-153-3965 Supplier Fritsch Milling And Sizing Inc Supplier No. 14.4430.32
Add to Cart
Add to Cart
Request a Quote
Impact rotor for PULVERISETTE 14 premium line with 6 ribs and cooling fins, pure titanium, fast comminution of fibrous substances - feed size < 15 mm.