Learn More
Fritsch Milling And Sizing Inc Rotor 6 Ribs, Stainless Steel PULVERISETTE 14 Premium Line
SDP
Catalog No. 501533949
Encompass
The 6-ribs impact rotor for PULVERISETTE 14 premium line with cooling fins is suitable for fast comminution of fibrous substances with a feed size of < 15 mm.
Catalog No. 50-153-3949 Supplier Fritsch Milling And Sizing Inc Supplier No. 14.4330.10
The 6-ribs impact rotor for PULVERISETTE 14 premium line with cooling fins is suitable for fast comminution of fibrous substances with a feed size of < 15 mm.