Learn More
Beckman Coulter Beckman Coulter rotor assembly JA-10.100 (B77584) - Fixed-angle rotor, 6 x 100 mL, 10,200 rpm; for Avanti J-15/J-15R centrifuges
Catalog No. NC2462663
Encompass
Beckman Coulter rotor assembly JA-10.100 (MPN B77584) is a Bio-Certified, aluminum fixed-angle rotor engineered to hold six 100 mL bottles and deliver up to 10,200 rpm (11,420 x g) for high-force separations. It is optimized for density gradient and differential separations in benchtop centrifuges.
  • Fixed-angle rotor for six 100 mL bottles.
  • Maximum speed 10,200 rpm (maximum RCF 11,420 x g).
  • Aluminum construction with Bio-Certified finish.
  • Suitable for density gradient and differential separations.
  • Holds standard 100 mL centrifuge bottles.
Catalog No. NC2462663 Supplier Beckman Coulter Supplier No. B77584
Beckman Coulter rotor assembly JA-10.100 (MPN B77584) is a Bio-Certified, aluminum fixed-angle rotor engineered to hold six 100 mL bottles and deliver up to 10,200 rpm (11,420 x g) for high-force separations. It is optimized for density gradient and differential separations in benchtop centrifuges.
  • Fixed-angle rotor for six 100 mL bottles.
  • Maximum speed 10,200 rpm (maximum RCF 11,420 x g).
  • Aluminum construction with Bio-Certified finish.
  • Suitable for density gradient and differential separations.
  • Holds standard 100 mL centrifuge bottles.