Learn More
Beckman Coulter Beckman Coulter JS-4.750 Rotor Assembly (Swinging-Bucket) for Avanti J-15 Series - B77580
Catalog No. NC1813263
Encompass
The JS-4.750 rotor assembly (MPN B77580) is a high-performance, four-place swinging-bucket rotor designed for use with Beckman Coulter Avanti J-15 series centrifuges. It accepts a wide range of tube and bottle adapters and delivers up to 5,250 xg for rapid sedimentation of cells, large particles, and protein precipitates.
  • Four swinging-bucket positions with 3 L nominal capacity.
  • Maximum speed 4,750 rpm and maximum RCF 5,250 xg.
  • Accepts adapters for 1.5 mL to 750 mL tubes, bottles, and blood bags.
  • Available multi-well plate carriers for high-throughput formats.
  • Constructed from stainless steel and aluminum for durability.
  • Biosafe design; sold as a single unit.
This item is not returnable. View return policy
Catalog No. NC1813263 Supplier Beckman Coulter Supplier No. B77580
Add to Cart
Add to Cart
Request a Quote
The JS-4.750 rotor assembly (MPN B77580) is a high-performance, four-place swinging-bucket rotor designed for use with Beckman Coulter Avanti J-15 series centrifuges. It accepts a wide range of tube and bottle adapters and delivers up to 5,250 xg for rapid sedimentation of cells, large particles, and protein precipitates.
  • Four swinging-bucket positions with 3 L nominal capacity.
  • Maximum speed 4,750 rpm and maximum RCF 5,250 xg.
  • Accepts adapters for 1.5 mL to 750 mL tubes, bottles, and blood bags.
  • Available multi-well plate carriers for high-throughput formats.
  • Constructed from stainless steel and aluminum for durability.
  • Biosafe design; sold as a single unit.