Learn More
Beckman Coulter ROTOR CLEANING BRUSH
Catalog No. NC9941335
Encompass_Preferred
Rotor Cleaning Brush, For cleaning rotor cavities, 1/4 to 1/2 in. Diameter, Minimum order of three
Catalog No. NC9941335 Supplier Beckman Coulter Supplier No. 347404
Rotor Cleaning Brush, For cleaning rotor cavities, 1/4 to 1/2 in. Diameter, Minimum order of three