Learn More
Beckman Coulter Rotor Cleaning Kit, quantity of one
Catalog No. NC9767938
Encompass
Rotor Cleaning Kit, Contains two 946 mL bottles of Solution 555 Rotor Cleaning Concentrate, 339379 Rotor Cleaning Brush, and 339380 Rotor Cleaning Brush
Catalog No. NC9767938 Supplier Beckman Coulter Supplier No. 339558
Rotor Cleaning Kit, Contains two 946 mL bottles of Solution 555 Rotor Cleaning Concentrate, 339379 Rotor Cleaning Brush, and 339380 Rotor Cleaning Brush