Learn More
Fritsch Milling And Sizing Inc Rotor, P-19, with V-Cutting Edges And Fixed kNives, Chromium-Free Tool Steel
SDP
Catalog No. 501534477
Encompass
This rotor with V-cutting edges and fixed knives made of chromium-free tool steel grinds the sample gently through a pure cutting action.
Alternatively a rotor with straight cutting edges and fixed knives made of chromium-free tool steel is offered, which also grinds the samples gently with a pure cutting action, but can only be used for easily cut materials and therefore does not offer an as effective size reduction like the rotor with V-cutting edges.
The rotors made of chromium-free tool steel are especially recommended for the sample preparation according to RoHS and WEEE (for the verification of hexavalent chromium) and for the XRF-analysis.
For all other applications we recommend our cutting tool set made of tool steel and hardmetal tungsten carbide, since these are harder and therefore more resilient against abrasion.
Catalog No. 50-153-4477 Supplier Fritsch Milling And Sizing Inc Supplier No. 45.5112.00
Add to Cart
Add to Cart
Request a Quote
This rotor with V-cutting edges and fixed knives made of chromium-free tool steel grinds the sample gently through a pure cutting action.
Alternatively a rotor with straight cutting edges and fixed knives made of chromium-free tool steel is offered, which also grinds the samples gently with a pure cutting action, but can only be used for easily cut materials and therefore does not offer an as effective size reduction like the rotor with V-cutting edges.
The rotors made of chromium-free tool steel are especially recommended for the sample preparation according to RoHS and WEEE (for the verification of hexavalent chromium) and for the XRF-analysis.
For all other applications we recommend our cutting tool set made of tool steel and hardmetal tungsten carbide, since these are harder and therefore more resilient against abrasion.