Learn More
Trajan Scientific VICI Valco Rotor Valco 6 Port Injection (VAL.SSAC6W)
Catalog No. 50635053
Encompass
VICI Valco Rotor, 6 Port, 5K PSI, 0.40mm Ports, VICI Valco Part Number SSAC6W, For VICI Valco Valve DC6W-K, (NEW LPP PN: VAL.SSAC6W). Made in the USA.
Catalog No. 50-635-053 Supplier Trajan Scientific Supplier No. 100.LC26R
VICI Valco Rotor, 6 Port, 5K PSI, 0.40mm Ports, VICI Valco Part Number SSAC6W, For VICI Valco Valve DC6W-K, (NEW LPP PN: VAL.SSAC6W). Made in the USA.