Learn More
Trajan Scientific VALCO Cheminert Rotor
Catalog No. NC1819025
Encompass
ALCO Cheminert Rotor (Valcon E5) for 6 port valve; 0.25 mm ports, Max Pressure: 15K psi.
Catalog No. NC1819025 Supplier Trajan Scientific Supplier No. VAL.C7215R6
ALCO Cheminert Rotor (Valcon E5) for 6 port valve; 0.25 mm ports, Max Pressure: 15K psi.