Learn More
Fisherbrand™ Rotor Knob
Catalog No. 12006909 Shop All Fisherbrand Products
Change view
Click to view available options
For Use With (Equipment):
Fisherbrand Mini Centrifuge
Catalog No. For Use With (Equipment)
12-006-909 Fisherbrand Mini Centrifuge
1 options
Catalog No. 12-006-909
Add to Cart
Add to Cart
Remove old knob with counter-clockwise turns. Fisherbrand™ Rotor Knob will engage with clockwise turns.
  • For use with Mini-Centrifuge
Warranty and Services
5 Years
Specifications
Material Plastic
Type Rotor Knob
Capacity None
Max. RCF None
Max. Speed None
For Use With (Equipment) Fisherbrand Mini Centrifuge
Biocontainment No
Autoclavable Not Autoclavable