Learn More
Thomas Scientific Rotor lid with clasp for 300D
SDP
Catalog No. 501459058
Encompass_Preferred
Extremely Quiet Digital Display Quick Spin Feature 24 Place Rotor 13,300 RPM (16,300xg) Cold Room Safe Two Year Warranty The Denville 300D Digital Microcentrifuge is compact, easy to use, yet affordable and powerful enough to allow every workstation to be equipped with its own individual centrifuge. The 300D has a unique easy access rotor and multi-flow air cooling system that allows for cool running. Parameters are selected with two "smart" knobs and values are shown on the large, easy to read, LED displays. Speed can be set and displayed in rpm or g-force. For samples that require below ambient temperatures, the 300D may be used in a cold room. The 300D is supplied with s 24 place angle easy access rotor that can accommodate tube sizes of 1.5ml and 2.0ml microcentrifuge tubes. Additional adapters are available for 0.5ml, 0.4ml, and 0.2ml tubes. The rotor is equipped with a cover that easily "snaps" on and off. The 300D is covered under Denville's two year warr
Catalog No. 50-145-9058 Supplier Thomas Scientific Supplier No. 1159Z75
Extremely Quiet Digital Display Quick Spin Feature 24 Place Rotor 13,300 RPM (16,300xg) Cold Room Safe Two Year Warranty The Denville 300D Digital Microcentrifuge is compact, easy to use, yet affordable and powerful enough to allow every workstation to be equipped with its own individual centrifuge. The 300D has a unique easy access rotor and multi-flow air cooling system that allows for cool running. Parameters are selected with two "smart" knobs and values are shown on the large, easy to read, LED displays. Speed can be set and displayed in rpm or g-force. For samples that require below ambient temperatures, the 300D may be used in a cold room. The 300D is supplied with s 24 place angle easy access rotor that can accommodate tube sizes of 1.5ml and 2.0ml microcentrifuge tubes. Additional adapters are available for 0.5ml, 0.4ml, and 0.2ml tubes. The rotor is equipped with a cover that easily "snaps" on and off. The 300D is covered under Denville's two year warr