Learn More
Fritsch Milling And Sizing Inc Rotor P-14 Cl with 24 Ribs Stainless Steel
SDP
Catalog No. 501534459
Encompass
Rotor p-14 cl with 24 ribs stainless steel for the grinding of fine materials with  feed particle sizes < 5 mm with highest air throughput.
The following impact rotors made of stainless steel are available:
8-rib rotor for the fast grinding of fibrous substances
12-rib rotor: for the grinding of samples with feed particle sizes of < 10 mm
Catalog No. 50-153-4459 Supplier Fritsch Milling And Sizing Inc Supplier No. 44.4240.10
Rotor p-14 cl with 24 ribs stainless steel for the grinding of fine materials with  feed particle sizes < 5 mm with highest air throughput.
The following impact rotors made of stainless steel are available:
8-rib rotor for the fast grinding of fibrous substances
12-rib rotor: for the grinding of samples with feed particle sizes of < 10 mm