Learn More
Fritsch Milling And Sizing Inc P-15 Rotor with Rotating And Fixed kNives, Tool Steel
SDP
Catalog No. 501534012
Encompass
Rotor with rotating and fixed knives made of tool steel for PULVERISETTE 15 grinds the sample gentle with a pure cutting action and can be recommended for most applications, because of its hardness and top resistance against abrasion.
The rotor made of chromium-free tool steel is especially recommended for sample preparation according to RoHS and WEEE (for example for trace of hexavelent chrome) or XRF analysis.
Catalog No. 50-153-4012 Supplier Fritsch Milling And Sizing Inc Supplier No. 15.4603.00
Rotor with rotating and fixed knives made of tool steel for PULVERISETTE 15 grinds the sample gentle with a pure cutting action and can be recommended for most applications, because of its hardness and top resistance against abrasion.
The rotor made of chromium-free tool steel is especially recommended for sample preparation according to RoHS and WEEE (for example for trace of hexavelent chrome) or XRF analysis.