Learn More
Fritsch Milling And Sizing Inc Rotor P-19, with Straight Cutting Edges And Fixed kNives, Chromium-Free Steel
SDP
Catalog No. 501534475
Encompass
This rotor with straight cutting edges and fixed knives made of chromium-free tool steel grinds the sample gently through a cutting action, but can only be used though for easily cut materials
Alternatively, a rotor with V-cutting edges and fixed knives made of chromium-free steel is offered, which also grinds the samples with a cutting action, but considerably more effective.
The rotors made of chromium-free steel are especially recommended for the sample preparation according to RoHS and WEEE (for the evidence of hexavalent chromium) and for the XRF-analysis.
For all other applications, we recommend our cutting tool set made of tool steel and hardmetal tungsten carbide, since these are harder and therefore more resilient against abrasion.
Catalog No. 50-153-4475 Supplier Fritsch Milling And Sizing Inc Supplier No. 45.5012.00
Add to Cart
Add to Cart
Request a Quote
This rotor with straight cutting edges and fixed knives made of chromium-free tool steel grinds the sample gently through a cutting action, but can only be used though for easily cut materials
Alternatively, a rotor with V-cutting edges and fixed knives made of chromium-free steel is offered, which also grinds the samples with a cutting action, but considerably more effective.
The rotors made of chromium-free steel are especially recommended for the sample preparation according to RoHS and WEEE (for the evidence of hexavalent chromium) and for the XRF-analysis.
For all other applications, we recommend our cutting tool set made of tool steel and hardmetal tungsten carbide, since these are harder and therefore more resilient against abrasion.