Learn More
Fritsch Milling And Sizing Inc Rotor P-25 Cpl., kNives And V-Cutting Edges Chromium Free
SDP
Catalog No. 501534504
Encompass
Grinds the sample through a gentle cutting action.
The rotor made of chromium-free tool steel is especially recommended for sample preparation according to RoHS and WEEE ( for the evidence of hexavalent chromium) and for the XRF-analysis.
For all other applications we recommend our cutting tool sets made of hardmetal tungsten carbide and therefore more resilient against abrasion.
Catalog No. 50-153-4504 Supplier Fritsch Milling And Sizing Inc Supplier No. 45.7112.00
Add to Cart
Add to Cart
Request a Quote
Grinds the sample through a gentle cutting action.
The rotor made of chromium-free tool steel is especially recommended for sample preparation according to RoHS and WEEE ( for the evidence of hexavalent chromium) and for the XRF-analysis.
For all other applications we recommend our cutting tool sets made of hardmetal tungsten carbide and therefore more resilient against abrasion.