Learn More
Trajan Scientific VICI Valco Rotor Seal, For Valco Valve, DC6UWE, 6 Port, 2 Position, For UW type Valve, 0.75 mm Ports, 1/16" Fittings
Catalog No. 501090378
Encompass
VICI Valco Rotor, For VICI Valco Valve, DC6UWE, 6 Port, 2 Position, For UW type Valve, 0.75 mm Ports, 1/16" Fittings. Made in the USA.
Catalog No. 50-109-0378 Supplier Trajan Scientific Supplier No. SSAC6UWE
VICI Valco Rotor, For VICI Valco Valve, DC6UWE, 6 Port, 2 Position, For UW type Valve, 0.75 mm Ports, 1/16" Fittings. Made in the USA.