Learn More
Sciencix ROTOR SEAL, VESPEL, 2 GROOVE
SDP
Catalog No. NC1051070
Encompass_Preferred
Rotor Seal, Vespel, 2-Groove, 7750-040, Equivalent to Agilent 0100-1853
Catalog No. NC1051070 Supplier Sciencix Supplier No. CTS10459
Rotor Seal, Vespel, 2-Groove, 7750-040, Equivalent to Agilent 0100-1853