Learn More
Waters Corp Waters Rotor Seal, Vespel, Rheodyne 7725i
Catalog No. 50793662 Shop All Waters Chromatography Solutions Products
Encompass_Preferred
The rotor seal is a Vespel-type seal designed for the Rheodyne model 7125 or 7725i Sample Injector. It creates a flow path between adjacent stator ports and includes a Teflon sleeve where the needle port tube pushes against the outer edge with spring tension. This seal is crucial for the sample injector’s operation, enabling a flow path and preventing the need for constant flushing of the sample loop, unless performing trace analysis.
  • Vespel-type seal
  • Compatible with Rheodyne 7125 and 7725i sample injector
  • Constructed with an embedded Teflon sleeve
  • Creates a flow path between adjacent stator ports
  • One unit per package
  • Replace as required
Catalog No. 50-793-662 Supplier Waters Corp Supplier No. WAT055946
The rotor seal is a Vespel-type seal designed for the Rheodyne model 7125 or 7725i Sample Injector. It creates a flow path between adjacent stator ports and includes a Teflon sleeve where the needle port tube pushes against the outer edge with spring tension. This seal is crucial for the sample injector’s operation, enabling a flow path and preventing the need for constant flushing of the sample loop, unless performing trace analysis.
  • Vespel-type seal
  • Compatible with Rheodyne 7125 and 7725i sample injector
  • Constructed with an embedded Teflon sleeve
  • Creates a flow path between adjacent stator ports
  • One unit per package
  • Replace as required