Learn More
Medchemexpress LLC HY-18980 10mg Medchemexpress, Rottlerin CAS:82-08-6 Purity:>98%
SDP
Catalog No. 501932825
Encompass_Preferred
Medchemexpress, HY-18980 10mg Rottlerin CAS:82-08-6 Rottlerin, a natural product purified from Mallotus Philippinensis, is a specific PKC inhibitor, with IC 50 values for PKCδ of 3-6 μM, PKCα,β,γ of 30-42 μM, PKCε,η,ζ of 80-100 μM. Rottlerin acts as a direct mitochondrial uncoupler, and stimulates autophagy by targeting a signaling cascade upstream of mTORC1. Rottlerin induces apoptosis via caspase 3 activation [1] [2] [3] . Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-193-2825 Supplier Medchemexpress LLC Supplier No. HY1898010MG
Medchemexpress, HY-18980 10mg Rottlerin CAS:82-08-6 Rottlerin, a natural product purified from Mallotus Philippinensis, is a specific PKC inhibitor, with IC 50 values for PKCδ of 3-6 μM, PKCα,β,γ of 30-42 μM, PKCε,η,ζ of 80-100 μM. Rottlerin acts as a direct mitochondrial uncoupler, and stimulates autophagy by targeting a signaling cascade upstream of mTORC1. Rottlerin induces apoptosis via caspase 3 activation [1] [2] [3] . Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.