Learn More
TARGETMOL CHEMICALS INC ROTTLERIN 25MG
SDP
Catalog No. 502577325
Encompass_Preferred
Also available in 5 mg 10 mg and bulk. Please contact Fisher for quotes. Rottlerin (NSC-56346) is a natural product purified from Mallotus Philippinensis and is a specific PKC inhibitor (IC50 PKC(delta) of 3-6 uM PKC(alpha)(beta)(gamma) of 30-42 uM PKC(epsilon)(eta)(zeta) of 80-100 uM). Rottlerin causes apoptosis via caspase 3 activation. purity: 97%
Catalog No. 50-257-7325 Supplier TARGETMOL CHEMICALS INC Supplier No. T1679125MG
Also available in 5 mg 10 mg and bulk. Please contact Fisher for quotes. Rottlerin (NSC-56346) is a natural product purified from Mallotus Philippinensis and is a specific PKC inhibitor (IC50 PKC(delta) of 3-6 uM PKC(alpha)(beta)(gamma) of 30-42 uM PKC(epsilon)(eta)(zeta) of 80-100 uM). Rottlerin causes apoptosis via caspase 3 activation. purity: 97%