Learn More
Abcam Rottlerin, Kinase inhibitor, 50MG
Catalog No. 502707636
Encompass_Preferred
MW 516.5 Da, Purity >95%. Originally reported to inhibit PKC isoforms, especially PKCδ and CAM-KIII. Also inhibits MAPKAP-K2 and PRAK (IC₅₀ values are 5.4 and 1.9 μM, respectively), CHK2, PLK1, PIM3 and SRPK1.
The product is subject to the following: Abcam Restricted Use Statement
Catalog No. 50-270-7636 Supplier Abcam Supplier No. AB12037750MG
MW 516.5 Da, Purity >95%. Originally reported to inhibit PKC isoforms, especially PKCδ and CAM-KIII. Also inhibits MAPKAP-K2 and PRAK (IC₅₀ values are 5.4 and 1.9 μM, respectively), CHK2, PLK1, PIM3 and SRPK1.
The product is subject to the following: Abcam Restricted Use Statement