Learn More
MSC AbilityOne SKILCRAFT Wet Mop Head, Looped End, Clamp Jaw, Medium, White
Catalog No. 502837836
Encompass
This mop head features a looped-end design and is constructed from a blend of cotton and polyester fleece, making it suitable for cleaning rough or textured surfaces like tile and concrete floors. It features a clamp jaw connection for secure attachment to most standard mop handles and is washable for easy maintenance.
  • Looped-end mop head for effective cleaning
  • Constructed from cotton and polyester fleece
  • Designed for rough or textured surfaces
  • Medium mop head size
  • Clamp jaw connection type
  • Washable for easy maintenance
  • Features a green vinyl headband with preshrunk tail band
  • White mop head color
Catalog No. 50-283-7836 Supplier MSC Supplier No. 53417275
This mop head features a looped-end design and is constructed from a blend of cotton and polyester fleece, making it suitable for cleaning rough or textured surfaces like tile and concrete floors. It features a clamp jaw connection for secure attachment to most standard mop handles and is washable for easy maintenance.
  • Looped-end mop head for effective cleaning
  • Constructed from cotton and polyester fleece
  • Designed for rough or textured surfaces
  • Medium mop head size
  • Clamp jaw connection type
  • Washable for easy maintenance
  • Features a green vinyl headband with preshrunk tail band
  • White mop head color