Learn More
MSC PIG SpillBlocker Dike, Polyurethane, Rough Surface, 10 ft., 1/Pk.
Catalog No. 5000684180
Encompass
This dike is designed for rapid emergency spill response on asphalt and other rough surfaces. Its construction features a tear-resistant top layer and a super-tacky bottom layer that conforms to surface irregularities, filling in cracks to establish a liquid-tight, secure seal. The unit confines and diverts hazardous or non-hazardous fluids without absorption, simplifying cleanup and reclamation efforts. It remains effective within a temperature range of 0°F to 160°F and can withstand liquid exposure up to 225°F for a maximum of 30 minutes.
  • Provides a liquid-proof seal on rough or uneven surfaces
  • Contains spills up to 2 in. deep
  • Resists water, oils, and many chemicals
  • Features high-visibility yellow color for improved safety
  • Includes a connector piece for joining multiple sections
  • Intended for one-time, limited-term use
Catalog No. 50-006-84180 Supplier MSC Supplier No. 38140844
This dike is designed for rapid emergency spill response on asphalt and other rough surfaces. Its construction features a tear-resistant top layer and a super-tacky bottom layer that conforms to surface irregularities, filling in cracks to establish a liquid-tight, secure seal. The unit confines and diverts hazardous or non-hazardous fluids without absorption, simplifying cleanup and reclamation efforts. It remains effective within a temperature range of 0°F to 160°F and can withstand liquid exposure up to 225°F for a maximum of 30 minutes.
  • Provides a liquid-proof seal on rough or uneven surfaces
  • Contains spills up to 2 in. deep
  • Resists water, oils, and many chemicals
  • Features high-visibility yellow color for improved safety
  • Includes a connector piece for joining multiple sections
  • Intended for one-time, limited-term use