Learn More
MSC SPI Surface Finish Comparator, 0.8 to 100 Ra μm, Nickel, 8 Specimens, 1 ea
Catalog No. 5000516814
Encompass
This surface finish comparator provides a reliable, portable solution for assessing surface roughness by sight and touch. Designed for machinists, inspectors, and designers, it contains eight specimens representing specific surface finishes, enabling accurate comparisons against machined parts.
  • Features eight individual specimens for shaping or planing assessment
  • Constructed from solid electroformed nickel for durability
  • Includes a protective case with instruction cards for field use
  • Marked in AA (micro inches), Ra (μm), Rz, and N numbers for versatility
  • Conforms to SAE, ANSI-B-46.1-1978, and military specifications
  • Individual specimen dimensions: 1.59 cm x 2.54 cm (5/8 in. x 1 in.)
Catalog No. 50-005-16814 Supplier MSC Supplier No. 06558258
This surface finish comparator provides a reliable, portable solution for assessing surface roughness by sight and touch. Designed for machinists, inspectors, and designers, it contains eight specimens representing specific surface finishes, enabling accurate comparisons against machined parts.
  • Features eight individual specimens for shaping or planing assessment
  • Constructed from solid electroformed nickel for durability
  • Includes a protective case with instruction cards for field use
  • Marked in AA (micro inches), Ra (μm), Rz, and N numbers for versatility
  • Conforms to SAE, ANSI-B-46.1-1978, and military specifications
  • Individual specimen dimensions: 1.59 cm x 2.54 cm (5/8 in. x 1 in.)