Learn More
Grainger Eagle Belting Round Belt, Polyurethane, 1/2 in. Dia., 95A, 100 ft., 1 ea.
Catalog No. 502758874
Encompass_Preferred
This solid, round polyurethane belt is designed for light to medium duty conveying applications, including packaging environments. Featuring a 95A durometer, it provides high elasticity and self-tensioning properties for reliable performance.
  • Diameter of 1/2 in.
  • Overall length of 100 ft. (30.5 m)
  • Temperature range of -22°F to 150°F (-30°C to 66°C)
  • Minimum pulley diameter of 5 in.
  • Smooth, non-reinforced construction
  • Abrasion, oil, and chemical-resistant
  • FDA approved for direct food contact
  • REACH and RoHS compliant
Catalog No. 50-275-8874 Supplier Grainger Supplier No. 15V498
This solid, round polyurethane belt is designed for light to medium duty conveying applications, including packaging environments. Featuring a 95A durometer, it provides high elasticity and self-tensioning properties for reliable performance.
  • Diameter of 1/2 in.
  • Overall length of 100 ft. (30.5 m)
  • Temperature range of -22°F to 150°F (-30°C to 66°C)
  • Minimum pulley diameter of 5 in.
  • Smooth, non-reinforced construction
  • Abrasion, oil, and chemical-resistant
  • FDA approved for direct food contact
  • REACH and RoHS compliant