Learn More
Grainger Eagle Belting Round Belt, Polyurethane, 1/8 in. Diameter, 85A, 100 ft.
Catalog No. 502758772
Encompass_Preferred
This solid-core, smooth round belt is engineered for reliable power transmission and conveying in demanding environments. Manufactured from orange polyurethane with a durometer of 85A, the belt provides a secure, weldable connection for closed-loop applications. Its durable material construction is designed to withstand oil, chemicals, and abrasion, making it well-suited for industrial processing tasks, including food packaging applications.
  • Features FDA-approved material for use in sensitive processing environments
  • Resistant to abrasion, oils, and chemicals for extended service life
  • Designed for a 1 in. minimum pulley diameter
  • Temperature operating range of -22°F to 150°F
  • Smooth surface finish facilitates efficient conveying
  • Available in a 100 ft. continuous length for custom cutting
Catalog No. 50-275-8772 Supplier Grainger Supplier No. 15V483
This solid-core, smooth round belt is engineered for reliable power transmission and conveying in demanding environments. Manufactured from orange polyurethane with a durometer of 85A, the belt provides a secure, weldable connection for closed-loop applications. Its durable material construction is designed to withstand oil, chemicals, and abrasion, making it well-suited for industrial processing tasks, including food packaging applications.
  • Features FDA-approved material for use in sensitive processing environments
  • Resistant to abrasion, oils, and chemicals for extended service life
  • Designed for a 1 in. minimum pulley diameter
  • Temperature operating range of -22°F to 150°F
  • Smooth surface finish facilitates efficient conveying
  • Available in a 100 ft. continuous length for custom cutting