Learn More
Grainger Eagle Belting Round Belt, Solid Polyurethane, 3/4 in., 100 ft.
Catalog No. 502759411
Encompass_Preferred
These round belts are engineered for reliable material transfer and light-duty power transmission in various applications, including packaging. Constructed from solid, non-reinforced polyurethane, they provide a durable solution that is self-tensioning and easy to weld on site. These belts feature a smooth surface and maintain performance across a broad temperature range.
  • Solid polyurethane construction with a smooth surface texture
  • FDA approved for safe operation in relevant environments
  • Excellent resistance to abrasion, oils, and chemicals
  • Self-tensioning design eliminates the need for take-ups
  • Operating temperature range from -30°C to 66°C (-22°F to 150°F)
  • Minimum pulley diameter of 7.5 in. (19.05 cm)
  • Closed loop belt with an overall length of 100 ft. (30.48 m)
Catalog No. 50-275-9411 Supplier Grainger Supplier No. 15V502
These round belts are engineered for reliable material transfer and light-duty power transmission in various applications, including packaging. Constructed from solid, non-reinforced polyurethane, they provide a durable solution that is self-tensioning and easy to weld on site. These belts feature a smooth surface and maintain performance across a broad temperature range.
  • Solid polyurethane construction with a smooth surface texture
  • FDA approved for safe operation in relevant environments
  • Excellent resistance to abrasion, oils, and chemicals
  • Self-tensioning design eliminates the need for take-ups
  • Operating temperature range from -30°C to 66°C (-22°F to 150°F)
  • Minimum pulley diameter of 7.5 in. (19.05 cm)
  • Closed loop belt with an overall length of 100 ft. (30.48 m)