Learn More
Grainger EAGLE BELTING Round Belt, Polyurethane, 5/16 in. Diameter, Solid, Orange, 100 ft.
Catalog No. 502760263
Encompass_Preferred
This solid round belt is designed for light to medium duty conveying applications. Constructed from high quality polyurethane, it features a smooth texture and is engineered for durability in demanding environments.
  • 85A hardness rating
  • FDA approved for food-safe applications
  • Resistant to abrasion, chemicals, and oils
  • Operating temperature range of -22°F to 150°F
  • Diameter of 5/16 in. and overall length of 100 ft.
  • Minimum pulley diameter of 2 1/2 in.
Catalog No. 50-276-0263 Supplier Grainger Supplier No. 15V486
This solid round belt is designed for light to medium duty conveying applications. Constructed from high quality polyurethane, it features a smooth texture and is engineered for durability in demanding environments.
  • 85A hardness rating
  • FDA approved for food-safe applications
  • Resistant to abrasion, chemicals, and oils
  • Operating temperature range of -22°F to 150°F
  • Diameter of 5/16 in. and overall length of 100 ft.
  • Minimum pulley diameter of 2 1/2 in.