Learn More
STOGGLES Round Black Fade Dimmers
Catalog No. NC2388221
Encompass_Preferred
Our Dimmers Lens are a UV-responsive lens that transition from light to dark based off of light exposure. Our light responsive lenses are anti-fog coated and shatter proof ensuring superior clarity and quality.
Catalog No. NC2388221 Supplier STOGGLES Supplier No. 162
Our Dimmers Lens are a UV-responsive lens that transition from light to dark based off of light exposure. Our light responsive lenses are anti-fog coated and shatter proof ensuring superior clarity and quality.