Learn More
Grainger Round Brush 3 In Dia
Catalog No. 502768084
Encompass_Preferred
The Advantech Round Brush is a 3 inch diameter round brush with crimped steel wire bristles and a threaded arbor hole. It has an overall length of 4 inches. This brush is designed for deburring, finishing, and surface conditioning applications. The crimped steel wire bristles provide a medium duty scrubbing action. With a max speed of 5,000 SFM, this brush can be used on metals, plastics, glass and wood. It fits in chucks, collets or adapters with a 1/4 inch internal thread. Made in the USA by Advantech.
Catalog No. 50-276-8084 Supplier Grainger Supplier No. 16Y897
The Advantech Round Brush is a 3 inch diameter round brush with crimped steel wire bristles and a threaded arbor hole. It has an overall length of 4 inches. This brush is designed for deburring, finishing, and surface conditioning applications. The crimped steel wire bristles provide a medium duty scrubbing action. With a max speed of 5,000 SFM, this brush can be used on metals, plastics, glass and wood. It fits in chucks, collets or adapters with a 1/4 inch internal thread. Made in the USA by Advantech.