Learn More
Grainger Round Brush Nylon 2 In Dia
Catalog No. 502767931
Encompass_Preferred
The Advantech Round Brush is a 2 inch diameter round brush with 0.014 inch diameter nylon bristles and a 5/8 inch trim length. It has a wood handle and is designed for light deburring, finishing, and polishing applications. This USA-made brush has a maximum operating speed of 15,000 RPM and temperature resistance up to 200°F. It features a 1/4 inch diameter arbor hole to fit power tools. The brush has an overall length of 2 inches.
Catalog No. 50-276-7931 Supplier Grainger Supplier No. 16Y896
The Advantech Round Brush is a 2 inch diameter round brush with 0.014 inch diameter nylon bristles and a 5/8 inch trim length. It has a wood handle and is designed for light deburring, finishing, and polishing applications. This USA-made brush has a maximum operating speed of 15,000 RPM and temperature resistance up to 200°F. It features a 1/4 inch diameter arbor hole to fit power tools. The brush has an overall length of 2 inches.