Learn More
Alkali Scientific Round Buckets, 2/pk
SDP
Catalog No. 502389135
Encompass_Preferred
Compatible with Z446 and Z446-K Universal High Capacity Centrifuges.
Catalog No. 50-238-9135 Supplier Alkali Scientific Supplier No. RS6376
Compatible with Z446 and Z446-K Universal High Capacity Centrifuges.