Learn More
Roboz Surgical ROUND CARBIDE BURR SIZE 1
SDP
Catalog No. NC0375032
Encompass
Carbide Round Burs; Size #1, 0.8mm Tip; 1.75 in. Length, 2.33mm Shaft Diameter; Pack Of 5 For use with the RS-6300 Ideal Micro Drill
Catalog No. NC0375032 Supplier Roboz Surgical Supplier No. RS6280C1
Carbide Round Burs; Size #1, 0.8mm Tip; 1.75 in. Length, 2.33mm Shaft Diameter; Pack Of 5 For use with the RS-6300 Ideal Micro Drill