Learn More
Roboz Surgical ROUND CARBIDE BURR SIZE 3
SDP
Catalog No. NC0375036
Encompass
Carbide Round Burs; Size #3, 1.2mm Tips; 1.75 in. Length; 2.33mm Shaft Diameter; Pack Of 5 For use with the RS-6300 Ideal Micro Drill
Catalog No. NC0375036 Supplier Roboz Surgical Supplier No. RS6280C3
Carbide Round Burs; Size #3, 1.2mm Tips; 1.75 in. Length; 2.33mm Shaft Diameter; Pack Of 5 For use with the RS-6300 Ideal Micro Drill