Learn More
Thomas Scientific Round cover glass, #1.5 thickness, 25 mm, 100 pack
SDP
Catalog No. NC1272770
Encompass_Preferred
High quality coverslips glasses are essential for microscopy and imaging. Cover glasses with diameters of 5 mm and 8 mm are now available from stock in convenient packages of 100.
Catalog No. NC1272770 Supplier Thomas Scientific Supplier No. 1217N82
High quality coverslips glasses are essential for microscopy and imaging. Cover glasses with diameters of 5 mm and 8 mm are now available from stock in convenient packages of 100.