Learn More
Alkali Scientific Round Cover Glass, Cover Slips for use within 12 Well TC Plates, 20mm Diameter, Thickness: 0.19-0.22mm, Pre-cleaned, 1 oz, 100/Sleeve
SDP
Catalog No. 502388775
Encompass_Preferred
ASI™ Round Cover Glass are high quality imported glass, and are non-cytotoxic. They are manufactured in 100,000 grade clean room and resistant to organic solvents and autoclaved by high temperature. Applicable to confocal experiment. Thickness: 0.19-0.22mm.
Catalog No. 50-238-8775 Supplier Alkali Scientific Supplier No. SM008
ASI™ Round Cover Glass are high quality imported glass, and are non-cytotoxic. They are manufactured in 100,000 grade clean room and resistant to organic solvents and autoclaved by high temperature. Applicable to confocal experiment. Thickness: 0.19-0.22mm.