Learn More
Alkali Scientific Round Cover Glass, Cover Slips for use within 6 Well TC Plates, 25mm Diameter, Thickness: 0.19-0.22mm, Pre-cleaned, 10 oz, 100/Sleeve
SDP
Catalog No. 502389351
Encompass_Preferred
ASI™ Round Cover Glass are high quality imported glass, and are non-cytotoxic. They are manufactured in 100,000 grade clean room and resistant to organic solvents and autoclaved by high temperature. Applicable to confocal experiment. Thickness: 0.19-0.22mm.
Catalog No. 50-238-9351 Supplier Alkali Scientific Supplier No. SM009
ASI™ Round Cover Glass are high quality imported glass, and are non-cytotoxic. They are manufactured in 100,000 grade clean room and resistant to organic solvents and autoclaved by high temperature. Applicable to confocal experiment. Thickness: 0.19-0.22mm.