Learn More
Thomas Scientific Small diameter round cover glass, #1 thickness, 3 mm
SDP
Catalog No. NC1629019
Encompass
High quality coverslips glasses are essential for microscopy and imaging. Cover glasses with diameters of 5 mm and 8 mm are now available from stock in convenient packages of 100. Available in #1.0 (0.15 mm), #15 (0.17 mm) and #20 (0.22 mm) thickness Available in round, square and rectangular geometries New 3 mm round for cranial window technique
Catalog No. NC1629019 Supplier Thomas Scientific Supplier No. 1217N66
High quality coverslips glasses are essential for microscopy and imaging. Cover glasses with diameters of 5 mm and 8 mm are now available from stock in convenient packages of 100. Available in #1.0 (0.15 mm), #15 (0.17 mm) and #20 (0.22 mm) thickness Available in round, square and rectangular geometries New 3 mm round for cranial window technique