Learn More
Thomas Scientific Round Cover Slips, German Glass 12mm
SDP
Catalog No. NC1418755
Encompass
Round Cover Slips, German Glass. Ideal for neural tissue cell cultures and hard-to-attach cells. Specially developed for fluorescence and confocal microscopy. #1 thickness.
This item is not returnable. View return policy
Catalog No. NC1418755 Supplier Thomas Scientific Supplier No. 1203J81
Round Cover Slips, German Glass. Ideal for neural tissue cell cultures and hard-to-attach cells. Specially developed for fluorescence and confocal microscopy. #1 thickness.