Learn More
Ted Pella Inc Coverslips, Glass, 12mm dia., 1 oz/pk
SDP
Catalog No. NC9194167
Encompass
Made from clear, high transmissive boro silicate glass. Cleaned, cut and packed in either 1 oz., 1/4 oz. or pkg/100. Available in various popular sizes ranging from Ø10 to Ø22mm. Not sterile.
Catalog No. NC9194167 Supplier Ted Pella Inc Supplier No. 26020
Made from clear, high transmissive boro silicate glass. Cleaned, cut and packed in either 1 oz., 1/4 oz. or pkg/100. Available in various popular sizes ranging from Ø10 to Ø22mm. Not sterile.