Learn More
Bellco Glass, Inc. Round Coverslip 25mm,#2
SDP
Catalog No. 501944706
Encompass_Preferred
Provides quality reliability and performance for all laboratory microscopy needs. Ideal for neutral cell cultures and hard to attach cells. Specially developed for fluorescence and con-focal microscopy. Size: 25mm od Thickness: #2 (.19-.25mm)
Catalog No. 50-194-4706 Supplier Bellco Glass, Inc. Supplier No. 194320002
Provides quality reliability and performance for all laboratory microscopy needs. Ideal for neutral cell cultures and hard to attach cells. Specially developed for fluorescence and con-focal microscopy. Size: 25mm od Thickness: #2 (.19-.25mm)