Learn More
Bellco Glass, Inc. Round Coverslip,5mm,#1 Thk
SDP
Catalog No. 501944701
Encompass_Preferred
Provides quality reliability and performance for all laboratory microscopy needs. Ideal for neutral cell cultures and hard to attach cells. Specially developed for fluorescence and con-focal microscopy. #1 thickness (0.13 to 0.16mm)
Catalog No. 50-194-4701 Supplier Bellco Glass, Inc. Supplier No. 194300005
Provides quality reliability and performance for all laboratory microscopy needs. Ideal for neutral cell cultures and hard to attach cells. Specially developed for fluorescence and con-focal microscopy. #1 thickness (0.13 to 0.16mm)