Learn More
Bellco Glass, Inc. RD German Coverslip, 12mm,#1
SDP
Catalog No. 501944702
Encompass_Preferred
Provides quality reliability and performance for all laboratory microscopy needs. Ideal for neutral cell cultures and hard to attach cells. Specially developed for fluorescence and con-focal microscopy. #1 thickness (0.13 to 0.16mm)
This item is not returnable. View return policy
Catalog No. 50-194-4702 Supplier Bellco Glass, Inc. Supplier No. 194310012A
Provides quality reliability and performance for all laboratory microscopy needs. Ideal for neutral cell cultures and hard to attach cells. Specially developed for fluorescence and con-focal microscopy. #1 thickness (0.13 to 0.16mm)