Learn More
Bellco Glass, Inc. RD German Coverslip, 25mm,#1
SDP
Catalog No. 501944704
Encompass_Preferred
Provides quality reliability and performance for all laboratory microscopy needs. Ideal for neutral cell cultures and hard to attach cells. Specially developed for fluorescence and con-focal microscopy. #1 thickness (0.13 to 0.16mm)
Catalog No. 50-194-4704 Supplier Bellco Glass, Inc. Supplier No. 194310025A
Provides quality reliability and performance for all laboratory microscopy needs. Ideal for neutral cell cultures and hard to attach cells. Specially developed for fluorescence and con-focal microscopy. #1 thickness (0.13 to 0.16mm)