Learn More
Sigma Aldrich Fine Chemicals Biosciences Round crystal window KBr d1EA
Catalog No. 502900644
Encompass
Very soft water soluble crystal low cost and good transmission range fogs.
Catalog No. 50-290-0644 Supplier Sigma Aldrich Fine Chemicals Biosciences Supplier No. Z1236091EA
Very soft water soluble crystal low cost and good transmission range fogs.